Disposable Face Shields
Disposable Face Shields
Actual product may vary
Fisher Catalogue
Disposable Face Shields
High-quality, optically clear, distortion-free 7mil film.
Have Questions?
Catalog NumberModel
FISS98127Full face
Catalog number FISS98127
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Model:
Full face
  • Polyester lens is treated with antifog and antistatic coating
  • Headpiece is held in place by a stretch strap and includes a latex-free foam band for comfortable extended wear
Specifications
Thickness (English)7 mil
Lens CoatingAnti-fog, Anti-static
Certifications/ComplianceOSHA 29 CFR 1910.1030, CDC
Lens TintClear
IncludesStretch Strap, Latex-free Foam Band
ModelFull face
SizeX-Large
MaterialPolyester
DisposableSingle-use
Lens MaterialPolyester
Unit SizePK