Blood Typing Kits with Simulated Blood
Actual product may vary
Innovating Science
Blood Typing Kits with Simulated Blood
Determine the ABO or ABO/Rh blood type of four unknown samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Application)
FISS96792RInnovating Science RefillSimulated abo/rh blood typing
FISS96792Blood Type KitSimulated abo/rh blood typing
Catalog number FISS96792R
Price (SGD)
-
Product Type:
Innovating Science Refill
For Use With (Application):
Simulated abo/rh blood typing
Use hands-on activities to understand the nature and importance of antigen-antibody reactions.
  • Provide accurate simulation of the blood typing procedure
  • No toothpicks, stirring or waiting for results
  • Kit contains materials for 10 groups
Kits use simulated blood only.
Specifications
Certifications/Compliance49 CFR 173.4
ColorWhite
Grade7 to 14
Age13+ years
For Use With (Application)Simulated abo/rh blood typing
IncludesInstructions, msds packet
Quantity1
Class Size10 Lab Groups
Product TypeInnovating Science Refill
Unit SizeEach