Spinbar™ Magnetic Stirring Bar Restrainer
Actual product may vary
Bel-Art
Spinbar™ Magnetic Stirring Bar Restrainer
Fits ergonomically in one hand and when placed alongside the outer wall of a mixing vessel, it draws in and secures the stirring bar in situ.
Have Questions?
Catalog NumberLength (Metric)
FISS967208 cm
Catalog number FISS96720
Price (SGD)
-
Length (Metric):
8 cm
  • Eliminate hazard caused with stir bar splashes when decanting mixing vessels
  • Comfortable, one-handed use
  • Fabricated with simple-to-clean durable polystyrene
  • Houses strong magnet that restrains magnetic stirring bars up to 80 mm (3 in.) long
Specifications
DescriptionFabricated with simple-to-clean durable polypropylene
Dimensions (L x W x H)8.71 x 2.87 x 4.24 cm
For Use With (Application)Eliminate hazard caused with stir bar splashes when decanting mixing vessels, capture bars in containers and restrains bars up to 3 in. long
For Use With (Equipment)Stirring bars
Length (English)3 in.
Diameter (Metric)8.71 cm
Length (Metric)8 cm
MaterialPolystyrene, Magnetic Alloy
Quantity1
ShapeRectangle
TypeStir Bar Retriever
Unit SizeEach