Forensic Chemistry of Blood Types Kit
Forensic Chemistry of Blood Types Kit
Actual product may vary
Forensic Chemistry of Blood Types Kit
Classify blood based on the presence or absence of erythrocytes on the surface of red blood cells.
Have Questions?
Catalog NumberProduct Type
FISS96644Forensic Chemistry of Blood Types Kit
Catalog number FISS96644
Price (SGD)
-
Product Type:
Forensic Chemistry of Blood Types Kit
Blood type, an inherited characteristic, is valuable to know in that it affects medical procedures, such as surgery and transfusions, paternity testing, as well as serving as evidence in criminal investigations.
Uses Innovating Science Simulated Blood; safe for classroom use.
Specifications
Certifications/Compliance49 CFR 173.4
Product TypeForensic Chemistry of Blood Types Kit
ColorWhite
Grade7 to 14
Age13+ years
For Use With (Application)For forensic chemistry of blood typing
IncludesBlood typing trays, 4 simulated blood samples, crime scene, 1 set abo/rh blood typing anti-sera, 1 pkg mixing stick, Teacher's manual and student study guide copymasters
Quantity1
Class Size15 Lab Groups
Unit SizeEA