Simple Bunsen Burner
Actual product may vary
Simple Bunsen Burner
Choice of LPG or natural gas burner.
Have Questions?
Catalog NumberGas Type
FISS95940LPG
Catalog number FISS95940
Price (SGD)
-
Gas Type:
LPG
  • Nickel-plated burner tube with air regulator and cylindrical connector
  • Mounted on pressed-steel base
Specifications
Product TypeBunsen Burner
For Use With (Equipment)LPG
Gas TypeLPG
Length (English)6 in.
Length (Metric)15.24 cm
MaterialAluminum, Brass, Steel
AutoclavableAutoclavable
DescriptionNickel plated
Diameter (English)3.4 in.
Diameter (English) Outer Mixing Tube0.51 in.
Diameter (Metric)85 mm (Base)
Diameter (Metric) Outer Mixing Tube13 mm
Height (English)4.3 in.
Height (Metric)142 mm
IncludesBunsen Burner
Weight (English)0.4 lb.
Weight (Metric)181.43 g
Width (English)3.5 in.
Width (Metric)8.89 cm
Unit SizeEach