PoxyGrid™ Labware Drain Stand
Actual product may vary
Bel-Art
PoxyGrid™ Labware Drain Stand
Three rows of hairpin-loop supports (14 in all) hold labware with 18 mm I.D. necks or larger.
Have Questions?
Catalog NumberPositions
FISS9518326
Catalog number FISS95183
Price (SGD)
-
Positions:
26
  • Top row has 12 straight pins 4 in. (102 mm) long to accommodate smaller items
  • Polystyrene tray catches water as tubes drain
  • Rubber-tipped feet for nonskid use
  • Wire loops on the back permit wall mounting
  • Dimensions: 16.75 x 6.5 x 19 in. (43 x 17 x 48 cm)
Specifications
AutoclavableAutoclavable
ColorBlue
HoldsLab Glassware
MaterialPolystyrene
Positions26
ShapeTriangle
StackableNo
SterilityNon-sterile
For Use With (Equipment)Lab Glassware
Size430 x 170 x 480 mm
Length (Metric)170 mm
Width (English)16.75 in.
Height (English)19 in.
Depth (English)6.5 in.
Depth (Metric)17 cm
IncludesDrip Tray
Product TypeBenchtop Dryer
ShelvesNone
Height (Metric)48 cm
Array12 x 1, 5 x 2, 4 x 1
Width (Metric)43 cm
Unit SizeEach