▸
Funnels
▸
Separatory Funnels
▸
Separatory Funnel
Actual product may vary
Fisherbrand
Separatory Funnel
Separatory Funnels provide excellent clarity and strength.
Have Questions?
Catalog Number
Capacity (Metric)
FISS90334C
500 mL
Catalog number
FISS90334C
Price (SGD)
-
Capacity (Metric):
500 mL
These pear-shaped polypropylene separating funnels feature excellent clarity and strength, and include a leak-proof threaded screw cap
The polypropylene stopcock is fitted with a self-lubricating PTFE plug
Not graduated
Specifications
Diameter (Metric)Top
90 mm
Capacity (Metric)
500 mL
Material
Polypropylene
Color
Clear
Length (Metric) Stem
45 mm
Product Type
Pear-shaped Separatory Funnel