Polypropylene Coplin Staining Jar
Actual product may vary
Bel-Art
Polypropylene Coplin Staining Jar
Large opening for easy placement and removal of slides.
Have Questions?
Catalog NumberSlide Count
FISS901305 or 10
Catalog number FISS90130
Price (SGD)
-
Slide Count:
5 or 10
  • Stains 3 × 1 in. (75 × 25mm) slides vertically
  • Holds five single slides or 10 slides back to back
  • Screw cap reduces leaks and evaporation
Specifications:
  • Steam autoclavable to 121°C (250°F)
  • Overall height: 4.5 in. (114mm)
  • Base diameter: 2.38 in. (59.7mm)
Specifications
AutoclavableAutoclavable
Bottom ShapeRound
Capacity (Metric)322 cm3
ColorWhite
DisposableReusable
MaterialPolypropylene
ShapeCylindrical
SterilityNon-sterile
Surface TreatmentNon-treated
TypeStaining Jar
For Use With (Application)For easier placement and removal of slides
Includes53 mm Polypropylene Flat-Top Screw Cap
QuantityEach
Slide Count5 or 10
Height (Metric)11.4 cm
Height (English)4.5 in.
Diameter (English)2.375 in.
Diameter (Metric)5.96 cm
Unit SizeEach