Class A Hexagonal Base Glass Graduated Measuring Cylinder
Class A Hexagonal Base Glass Graduated Measuring Cylinder
Actual product may vary
Fisherbrand
Class A Hexagonal Base Glass Graduated Measuring Cylinder
The Class A Hexagonal Base Glass Graduated Measuring Cylinder features printed graduations and a spout.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsScale Color
FISS884551000 mL10 mLBlue
FISS8845150 mL1 mLBlue
FISS88452100 mL1 mLBlue
Catalog number FISS88455
Price (SGD)
-
Capacity (Metric):
1000 mL
Graduations:
10 mL
Scale Color:
Blue
  • Made from high quality Borosilicate 3.3 Glass
  • Hexagonal base for maximum stability
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class A
  • Conforms to DIN EN ISO 4788 standard
  • Calibrated to contain
  • Autoclavable
Specifications
Base TypeHexagonal
CalibrationCalibrated to contain
Diameter (Metric) Inside65 mm
Scale ColorBlue
ShapeRound
Certifications/ComplianceDIN EN ISO 4788
ClassA
Height (English)18 in.
Height (Metric)45 cm
Tolerance Metric±5 mL
TypeGraduated Measuring Cylinder
MaterialBorosilicate Glass
Diameter (Metric) Outside74 mm
Capacity (Metric)1000 mL
For Use With (Application)Measuring Liquids
Graduations10 mL