Check Order Status
Solid Glass Beads
Actual product may vary
Fisherbrand
Solid Glass Beads
Increase the efficiency of separations.
Have Questions?
Catalog Number
Diameter (Metric)
Quantity
FISS800242
5 mm
2700/Pk.
Catalog number
FISS800242
Price (SGD)
-
Diameter (Metric):
5 mm
Quantity:
2700/Pk.
For use in distillations or as inert boiling stones
Packaged in 1 lb. (0.45kg) packs
Specifications
Material
Glass
Diameter (Metric)
5 mm
Quantity
2700/Pk.