Check Order Status
Silica Gel Desiccant (6-14 Mesh, Certified ACS), Fisher Chemical™
Element Search
Structure Search
Silica Gel Desiccant (6-14 Mesh, Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FISS684-212
2.5 kg
Poly Bottle
Catalog number
FISS684-212
Price (SGD)
-
Quantity:
2.5 kg
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
2230°C
Chemical Name or Material
Silica Gel Desiccant
Color
White
Melting Point
1710°C
Physical Form
Solid
View more