Check Order Status
Disposable Microscope Slides
Actual product may vary
Disposable Microscope Slides
Durable plastic microscope slides.
Have Questions?
Change view
Catalog Number
Quantity
FISS67112A
144/Pk.
FISS67111A
12/Pk.
2 Options
Catalog number
FISS67112A
Price (SGD)
-
Quantity:
144/Pk.
Durable plastic microscope slides.
Precleaned and packed per dozen
Size: 3 × 1 in. (7.6 × 2.5cm)
Specifications
Length (Metric)
75 mm
Material
Plastic
Precleaned
Yes
Width (Metric)
25 mm
Quantity
144/Pk.