Alcohol Burner Stand
Actual product may vary
United Scientific Supplies
Alcohol Burner Stand
The Alcohol Burner Stand accommodates a variety of burner sizes.
Have Questions?
Catalog NumberIncludes
FISS66967Heavy Wire Gauze Top
Catalog number FISS66967
Price (SGD)
-
Includes:
Heavy Wire Gauze Top
  • Made of chrome-plated steel wire with heavy wire gauze top
  • Overall height is 5 in. (12.7 cm); accommodates all burner sizes
  • S67184 features a ceramic center on the wire gauze top
Specifications
Product TypeStand
For Use With (Equipment)Accomodates burners of all sizes
Gas TypeAll
Length (English)4 in.
Length (Metric)101.6 mm
MaterialPlated Steel
Size5 in. Tall
ConstructionSteel wire, Zinc-plated
Height (English)5 in.
Height (Metric)12.7 cm
IncludesHeavy Wire Gauze Top
Unit SizeEach