Check Order Status
Proguard™ Professional Handcream
Actual product may vary
Proguard™ Professional Handcream
Protects from prolonged glove use and frequent hand scrubbing.
Have Questions?
Catalog number
FISS66872
Price (SGD)
-
Unscented handcream protects from chapping and irritation
Particularly effective for improving latex glove comfort
Nonpetroleum, nongreasy and absorbs instantly
Latex-safe
Specifications
Product Type
Professional Hand Cream
Color
White
Form
Cream
For Use With (Application)
Protects from prolonged glove use and frequent hand scrubbing.
Packaging
Pump
Quantity
1 Ea.
Volume (English)
16 oz.
Volume (Metric)
473.1 mL