Faceshield
Faceshield
Actual product may vary
Faceshield
Comes apart for easy disinfection.
Have Questions?
Catalog number FISS66730
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Large, 8.5 in.H (21.6cm) molded window
  • 99.9% UV protection up to 380nm
  • Ledge on the headgear prevents fluid from splashing down inside the window
  • Pin-lock headgear can be sterilized and disinfected; window can be steam autoclaved
Specifications
Height (English)8.5 in.
Height (Metric)21.6 cm
Certifications/ComplianceMeets ANSI Z87.1-1989
Unit SizeEA