Check Order Status
Qualitative-Grade Filter Paper Circles
Actual product may vary
Fisherbrand
Qualitative-Grade Filter Paper Circles
Limited wet strength.
Have Questions?
Catalog Number
Grade
Diameter (Metric)
FISS47571C
P5
11 cm
Catalog number
FISS47571C
Price (SGD)
-
Grade:
P5
Diameter (Metric):
11 cm
Coarse Porosity
Flowrate: Fast, 175mL/min.
Surface: Smooth, open texture
Particle Retention: >20μm
Applications: Suitable for coarse and gelatinous precipitates
Medium Porosity
Flowrate: Medium, 60mL/min.
Surface: Smooth
Particle Retention: 5 to 10μm
Applications: General lab uses
Fine Porosity
Flowrate: Slow, 5mL/min.
Surface: Smooth, dense texture
Particle Retention: 1 to 3μm
Applications: Excellent retention; suitable for finest particles or precipitates
Specifications
Format
Disc
Particle Retention
5 to 10 μm
Filtration Speed
60mL/min.
For Use With (Application)
General lab use
Grade
P5
Porosity
Medium
Quantity
100/Pk.
Diameter (Metric)
11 cm