Steel Retort Base Stand and Rod Set
Actual product may vary
Steel Retort Base Stand and Rod Set
Steel Retort Base Stand and Rod Set
Steel Retort Base Stand and Rod Set
Steel Retort Base Stand and Rod Set
Eisco steel retort base stand and rod sets are premium quality metalware built to provide maximum stability through years of rugged lab applications.
Have Questions?
Catalog NumberDimensions (L x W)Length (Metric) Rod
FISS4171025 x 16 cm (Base)60 cm
Catalog number FISS41710
Price (SGD)
-
Dimensions (L x W):
25 x 16 cm (Base)
Length (Metric) Rod:
60 cm
  • Set includes (1) stamped steel retort base and (1) steel rod
  • Base is powder coated for chemical resistance, includes threaded hole in center of short end
  • Steel rod includes 10 x 1.5 mm threading
  • Screw together before use
Specifications
MaterialSteel
Height (Metric)5 cm
Height (English)2 in.
Dimensions (L x W)25 x 16 cm (Base)
Length (English) Rod23.6 in.
Length (Metric)59.9 cm
Length (Metric) Base25 cm
Length (Metric) Rod60 cm
Width (English)6 in.
Width (English) Base6.25 in.
Width (Metric)15.24 cm
Width (Metric) Base16 cm
Diameter (Metric) Rod12 mm
IncludesRod, base
Product TypeStands and Rings
Thread Size10 x 1.5 mm
AutoclavableAutoclavable
ColorBlue, Silver
DescriptionSturdy construction
For Use With (Application)Lab stand
Quantity1 Set
Weight (English)4 lb.
Weight (Metric)1.81 kg
Unit SizeEach