Solid Stainless Steel Hooked Weights
Solid Stainless Steel Hooked Weights
Actual product may vary
Fisherbrand
Solid Stainless Steel Hooked Weights
Solid stainless steel hooked weights with integrated hooks. Weights are cylindrical with natural stainless steel color, weight stamped at top
Have Questions?
Catalog NumberWeight (Metric)
FISS416081000 g
Catalog number FISS41608
Price (SGD)
-
Weight (Metric):
1000 g
  • Solid stainless steel construction
  • Hook is integrated with weight
  • Natural stainless steel color with stamped identifier at top
  • Weight is cylindrical
Note:
The Lengths indicated for each weight do not include the hook.
Specifications
Product TypeCylindrical Weight
MaterialStainless Steel
Weight (Metric)1000 g
Length (Metric)5 cm
For Use With (Application)Calibrating scales and balances
IncludesWeight with hook
Quantity1
Unit SizeEA