Electronic Digital Micrometer
Electronic Digital Micrometer
Actual product may vary
Electronic Digital Micrometer
Offers metric/inch conversion, origin setting, data hold, relative/absolute measuring and tolerance setting.
Have Questions?
Catalog NumberMaterial
FISS40502AStainless Steel
Catalog number FISS40502A
Price (SGD)
-
Material:
Stainless Steel
  • Measures from 0 to 25mm
  • Made of hardened and ground stainless steel
  • Resolution: 0.001mm
  • Accuracy: 0.002mm
  • RS-232 port connects micrometer to a printer or computer with user-supplied cable
Specifications
Product TypeDigital Micrometer
MaterialStainless Steel
For Use With (Application)Offers metric/inch conversion, origin setting, data hold, relative/absolute measuring and tolerance setting
Includes1.5V Battery and impact-resistant ABS Plastic case, spanner, Foam lined with Metal hinges and snap locked lacquered storage box, large digital display
Unit SizeEach