Disposable Aluminum Dishes
Disposable Aluminum Dishes
Actual product may vary
Disposable Aluminum Dishes
Withstand temperatures up to 677°C (1250.6°F).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Inner Top
FISS4028450 mL5.84 cm
FISS4028320 mL4.39 cm
Catalog number FISS40284
Price (SGD)
-
Capacity (Metric):
50 mL
Diameter (Metric) Inner Top:
5.84 cm
  • Designed for weighing, dispensing and storing
  • Can be used as evaporating dishes or as dust covers
  • With crimped sides for rigidity and a smooth bottom
Specifications
Bottom ShapeFlat
Capacity (Metric)50 mL
DisposableSingle-use
MaterialAluminum
ShapeRound
TypeWeighing Dish
For Use With (Application)Designed for weighing, dispensing and storing, can be used as evaporating dishes or as dust covers
Diameter (Metric) Inner Top5.84 cm
Diameter (Metric)5.84 cm (Inner Top)
Unit SizePK