Porcelain Evaporating Dish, Flat Form
Porcelain Evaporating Dish, Flat Form
Actual product may vary
Fisherbrand
Porcelain Evaporating Dish, Flat Form
Shallow form with lip and flat bottom. Glazed except for rim and part of outside bottom.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FISS39348100 mL100 mm25 mm
FISS3934650 mL75 mm20 mm
Catalog number FISS39348
Price (SGD)
-
Capacity (Metric):
100 mL
Diameter (Metric):
100 mm
Height (Metric):
25 mm
  • Shallow form with lip and flat bottom.
  • Glazed except for rim and part of the outside bottom.
  • Autoclavable.
  • Withstand temperatures to 1150°C.
  • To prevent thermal stress cracks, we strongly recommend a heating/cooling rate not exceeding 200°C per hour.
  • United porcelain evaporating dishes offer resistance to acids and alkalis except for hydrofluoric acid.
Specifications
AutoclavableAutoclavable
Bottom ShapeFlat
Capacity (Metric)100 mL
ColorWhite
MaterialPorcelain
ShapeRound
SterilityNon-sterile
Surface TreatmentGlaze
TypeEvaporating Dish
Volume (Metric) Working100 mL
Temperature Range (Metric)Up to 1150°C
Height (Metric)25 mm
Diameter (Metric)100 mm
Unit SizePK