Flow Indicator, SAN Body
Actual product may vary
Dynalon
Flow Indicator, SAN Body
Clear outer case on allows for unobstructed view
Have Questions?
Catalog NumberAirflow Rate
FISS371985 to 95 mL/sec.
Catalog number FISS37198
Price (SGD)
-
Airflow Rate:
5 to 95 mL/sec.
This SAN, bright red paddlewheel flow meter indicates liquid or gas flow from 5 to 95mL/second. The clear outer case on flow indicator allows for unobstructed view of the wheel which spins freely with flow. Paddlewheel flow indicator is not for use with corrosive fluids or temperature over (185°F) 85°C. Used to connect tubing I.D. of 3/8 in. to 9/16 in. (10-14mm) I.D.
  • For liquid flows down to 5mL/second
  • Wheel spins freely and speed of rotation is proportional to flow rate
  • Maximum recommended flow rate up to 95mL/sec
• Maximum 2 bar pressure• Not to be used with corrosive fluids or over +85°C• Can be used with gases For liquid flows down to 5 ml/second Wheel spins freely and speed of rotation is proportional to flow rate Maximum recommended flow rate up to 95 ml/sec Maximum 2 bar pressure Not to be used with corrosive fluids or over +85°C Can be used with gases
Specifications
TypePaddle Wheel Flow Indicator
Dimensions (L x W x H)3.53 x 0.62 x 1.6 in.
Airflow Rate5 to 95 mL/sec.
Waterflow Rate5 to 95 mL/sec.
Unit SizeCase