Rubber Bulbs, Natural
Rubber Bulbs, Natural
Actual product may vary
Fisherbrand
Rubber Bulbs, Natural
Replacement Bulbs for dropping bottles, medicine droppers and small pipets.
Have Questions?
Catalog NumberVolume (Metric)
FISS323252 mL
Catalog number FISS32325
Price (SGD)
-
Volume (Metric):
2 mL
Replacement bulbs for dropping bottles, medicine droppers and small pipets.
  • Made of natural rubber, natural color
  • 2 mL capacity, 52 mm L x 13 mm OD (6 mm hole dia.)
  • 1 mL capacity, 43 mm L x 13 mm OD (5 mm hole dia.)
  • Designed for use with Pasteur pipettes, small pipettes, and medicine droppers
Specifications
Compatible Tips2 mL
AutoclavableNot Autoclavable
ColorNatural
DisposableReusable
For Use WithDropping Bottle, Medicine Dropper, Small Pipette
Grip TypeSingle-handed
MaterialRubber
Model52mmL x 13mmOD
Number of Channels1
SterilityNon-sterile
TypeBulb
Volume (Metric)2 mL
For Use With (Equipment)Dropping bottles, medicine droppers and small pipets
Length (English)2 in.
Length (Metric)52 mm
Diameter (English) Outer0.5 in.
Diameter (Metric) Outer13 mm
Unit SizePK