Round Lead Ring Flask Weights with Vikem™ Vinyl Coating
Actual product may vary
Bel-Art
Round Lead Ring Flask Weights with Vikem™ Vinyl Coating
Protects and stabilizes expensive glassware. Round Lead Ring Flask Weights with Vikem™ Vinyl Coating reduces floating, breakage and chemical spills.
Have Questions?
Catalog NumberDiameter (English) InnerDiameter (English) OuterWeight (English)
FISS31719B2 in.3.5 in.1 lb.
Catalog number FISS31719B
Price (SGD)
-
Diameter (English) Inner:
2 in.
Diameter (English) Outer:
3.5 in.
Weight (English):
1 lb.
  • Stabilize glassware immersed in water baths; reduces floating, breakage and chemical spills
  • Bright red Vikem vinyl coating
  • Chemical-resistant
  • Will not scratch flasks
Specifications
AutoclavableNot Autoclavable
ColorRed
HoldsFlasks (250 to 1000 mL)
MaterialVinyl-coated Lead
Positions1
ShapeRing
StackableYes
SterilityNon-sterile
For Use With (Equipment)Erlenmeyer-style straight-necked flasks
Length (Metric)89 mm
Diameter (English) Inner2 in.
Diameter (English) Outer3.5 in.
Diameter (Metric) Outer8.9 cm
Weight (English)1 lb.
Weight (Metric)454 g
Product TypeLead Ring Flask Weight
Diameter (Metric) Inner5.1 cm
ShelvesNone
Height (Metric)38 mm
Width (Metric)89 mm
Unit SizeEach