Plastic Beaker with Handle
Actual product may vary
Dynalon
Plastic Beaker with Handle
Our handles make your handling of solutions easier and safer.
Have Questions?
Catalog NumberCapacity (Metric)
FISS307285000 mL
Catalog number FISS30728
Price (SGD)
-
Capacity (Metric):
5000 mL
  • Durable beakers feature a low form design for better stability
  • Made of autoclavable polypropylene
Specifications
ColorTranslucent
Diameter (English)Top7.5 in.
Diameter (Metric)Top19 cm
Capacity (Metric)5000 mL
MaterialPolypropylene
Graduation Range250 and 5000 mL
Height (English)8.9 in.
Height (Metric)22.6 cm
IncludesHandle
TypeBeaker