Specifications
DescriptionGlass Stoppers
Diameter (Metric)2 cm
For Use With (Application)Seal flasks
Height (Metric)3.6 cm
Length (Metric)2 cm
Quantity1
Size24/29
Standard Taper Joint19/26
Width (Metric)2 cm
AutoclavableAutoclavable
ColorClear
Diameter (Metric) Outer19 mm
MaterialBorosilicate Glass
Product TypePennyhead Stopper
ShapeCylindrical
Unit SizeEach