Dual Sided Microtube Rack with Lid
Actual product may vary
Dual Sided Microtube Rack with Lid
Eisco™ Dual Sided Microtube Rack with Lid allows good visibility of contents and alpha-numeric print on the rack for indexing.
Have Questions?
Catalog NumberPositions
FISS29413108
Catalog number FISS29413
Price (SGD)
-
Positions:
108
  • Rugged polypropylene dual sided storage rack with lid
  • One side holds 108 x 0.5 mL microtubes
  • Second side holds 108 x 1.5 mL microtubes
  • Letter and numbers on surface for easy indexing
  • Transparent lid for easy visibility
  • 8 x 5.25 x 2 in. outer dimensions with lid on
  • Rack and lid are both autoclavable
Specifications
Closure IncludedYes
AutoclavableAutoclavable
ColorAssorted
HoldsMicrocentrifuge Tubes (0.5 mL)
MaterialPolypropylene
Positions108
ShapeRectangular
StackableYes, When Empty
SterilityNon-sterile
For Use With (Application)Storing Microtubes
For Use With (Equipment)Microtubes
Quantity1
Length (Metric)20 cm
IncludesLid
Product TypeMicrotube Rack
ShelvesNone
Height (Metric)5 cm
Array12 x 9
Width (Metric)13.3 cm
Unit SizeEach