Polypropylene Filtering Flasks
Polypropylene Filtering Flasks
Actual product may vary
Fisherbrand
Polypropylene Filtering Flasks
Buchner filtering flask made of high quality, chemical resistant polypropylene with a conical shape to prevent spills
Have Questions?
Catalog NumberCapacity (Metric)
FISS270611000 mL
Catalog number FISS27061
Price (SGD)
-
Capacity (Metric):
1000 mL
  • Made of high quality, chemical resistant polypropylene
  • Conical shape, beaded rim to prevent drips
  • Autoclavable at 121°C
  • Microwavable
  • Ungraduated
Specifications
Capacity (Metric)1000 mL
MaterialPolypropylene
Neck StyleBeaded Rim
Product TypeFiltering Flask
ShapeConical
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
Mouth StyleNarrow
SterilityNon-sterile
Bottom ShapeFlat
Unit SizeEA