Check Order Status
Vinegar
Element Search
Structure Search
Vinegar
Have Questions?
Catalog Number
Quantity
Packaging
FISS25623
500 mL
Liquid - White
Catalog number
FISS25623
Price (SGD)
-
Quantity:
500 mL
Packaging:
Liquid - White
Request bulk or custom format
Specifications
Chemical Name or Material
Vinegar
Shelf Life
5 years
CAS
64-19-7
ChemAlert Storage Symbol
Gray
Color
Colorless
View more