Check Order Status
Hydrochloric Acid, ACS, 12M
Element Search
Structure Search
Fisherbrand
Hydrochloric Acid, ACS, 12M
Have Questions?
Catalog Number
Quantity
Packaging
FISS25358
500 mL
ACS Grade - SAFE-COTE
Catalog number
FISS25358
Price (SGD)
-
Quantity:
500 mL
Packaging:
ACS Grade - SAFE-COTE
Request bulk or custom format
Specifications
Chemical Name or Material
Hydrochloric Acid
Shelf Life
2 years
Color
Colorless
CAS
7647-01-0
Density
1.18
View more