Bromothymol Blue, Sodium Salt
Bromothymol Blue, Sodium Salt
Bromothymol Blue, Sodium Salt
Have Questions?
Catalog NumberQuantityPackaging
FISS252065 gReagent Grade
Catalog number FISS25206
Price (SGD)
-
Quantity:
5 g
Packaging:
Reagent Grade
Request bulk or custom format
Specifications
Chemical Name or MaterialBromothymol Blue, Sodium Salt
Shelf Life5 years
CAS34722-90-2
ChemAlert Storage SymbolGray
Assay Percent Range100% w/v
View more
CO2/pH indicator