Class A Glass Graduated Cylinders, ASTM
Class A Glass Graduated Cylinders, ASTM
Actual product may vary
Class A Glass Graduated Cylinders, ASTM
Eisco™ Glass Graduated Cylinders feature a hexagonal base, printed graduations, and spout.
Have Questions?
Catalog NumberCapacity (Metric)Scale Color
FISS245081000 mLBlue
Catalog number FISS24508
Price (SGD)
-
Capacity (Metric):
1000 mL
Scale Color:
Blue
  • Made from high quality borosilicate 3.3 glass
  • Hexagonal base for extra stability
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class A
  • Conforms to ASTM E1272 standard
  • Autoclavable
Specifications
Base TypeHexagonal
Diameter (Metric) Inside63 mm
Scale ColorBlue
ShapeRound
Certifications/ComplianceASTM E1272
ClassClass A
Height (English)18 in.
Tolerance Metric±3 mL
TypeGraduated Measuring Cylinder
MaterialBorosilicate Glass
Diameter (Metric) Outside75 mm
Capacity (Metric)1000 mL
Graduations10 mL
AutoclavableAutoclavable
Capacity1000mL
Subdivision10mL
Unit SizeEA