Polypropylene Measuring Jug, Short Form Euro Design
Polypropylene Measuring Jug, Short Form Euro Design
Actual product may vary
Polypropylene Measuring Jug, Short Form Euro Design
Polypropylene Measuring Jug, Short Form Euro Design
Polypropylene Measuring Jug, Short Form Euro Design
Fisherbrand
Polypropylene Measuring Jug, Short Form Euro Design
Graduated measuring jugs are constructed of premium quality polypropylene plastic, short form design featuring a handle and elongated spout for easy pouring.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISS238013000 mL
FISS238025000 mL
FISS238002000 mL
Catalog number FISS23801
Price (SGD)
-
Capacity (Metric):
3000 mL
  • Constructed of premium quality polypropylene plastic
  • Optically clear with easy to read, raised graduations
  • Short form (Euro) design with elongated tapered spout for easy pouring and transferring of liquids
  • Handle allows for safe and convenient lifting
  • Capable of withstanding temperatures up to 135°C
  • Resistant to strong acids, alcohols, and bases
  • Autoclavable, microwavable, and dishwasher safe
Specifications
Capacity (Metric)3000 mL
Capacity (English)101.44 oz
ColorTranslucent
FormShort Form
IncludesHandle, Tapered Spout
MaterialPolypropylene
Max. Temperature (Metric)135°C
SterilityNon-sterile
Graduations100 mL
Closure IncludedNo
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
MouthExtra Wide
ShapeCylindrical
Product TypeMeasuring Jug
Unit SizeEA