▸
Mixers
▸
Mortars and Pestles
▸
Mortar and Pestle Set
Actual product may vary
Mortar and Pestle Set
Features a flat bottom, unglazed grinding surfaces, and spout for easy pouring. Eisco™ Mortar and Pestle Set is made from lab quality porcelain and designed for efficient and uniform grinding of various materials.
Have Questions?
Catalog Number
Capacity (Metric)
Diameter (Metric) Inner
FISS23643
550 mL
153 mm
Catalog number
FISS23643
Price (SGD)
-
Capacity (Metric):
550 mL
Diameter (Metric) Inner:
153 mm
Made from lab quality porcelain
Outside of mortar and handle of pestle is glazed
Inner bowl and pestle end are not glazed
Spout for easing pouring
Flat bottom for stability
Specifications
Capacity (Metric)
550 mL
Finish
Unglazed
Capacity (English)
18.5 oz.
Diameter (English) Outer
6 in.
Diameter (Metric) Inner
153 mm
Product Type
Mortar and Pestle
Material
Porcelain
Capacity
550 ml
Type
Mortars and Pestles