Immersion Oil
Actual product may vary
Cargille
Immersion Oil
Finest immersion oil available.
Have Questions?
Catalog NumberTypeViscosityQuantity
FISS19305Immersion Oil Type A150 cSt4 oz.
Catalog number FISS19305
Price (SGD)
-
Type:
Immersion Oil Type A
Viscosity:
150 cSt
Quantity:
4 oz.
For use with 100X oil immersion objectives. Type B is recommended for most educational microscopy; contains no PCB compounds.
  • Type A: Low fluorescence, low viscosity
  • Type B: Low fluorescence, high viscosity
Specifications
TypeImmersion Oil Type A
Viscosity150 cSt
Certifications/ComplianceISO 8036/1
Quantity4 oz.
Refractive Index1.5150 ±0.0002
Unit SizeEach