Soil Test Kit
Soil Test Kit
Actual product may vary
Soil Test Kit
Accurate test results within the hour.
Have Questions?
Catalog number FISS17702
Price (SGD)
-
  • Measure soil's pH, nitrogen, phosphorus and potassium — up to 10 times each
  • Color comparator system
  • Color-coded reagent capsules perform 40 total tests
Specifications
Product TypeSoil Test Kit
For Use With (Application)Measure the soil ph, nitrogen, phosphorous and potassium
IncludesEasy-to-use capsule system, 4 individual color comparators, ph and npk levels chart and complete instructions
Time Range1
Dimensions (L x W x H)W: 9.13 in.; H: 8.5 in.; D: 1.31 in.
Unit SizeEA