Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™
Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™
Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™
Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FISS162-2122.5 kgPoly Bottle
Catalog number FISS162-212
Price (SGD)
-
Quantity:
2.5 kg
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS112926-00-8 , 7646-79-9
Specifications
Chemical Name or MaterialSilica Gel Desiccant, Indicating
ColorBlue
Physical FormSolid
Name Note4-10 Mesh, Grade 48
CAS112926-00-8
View more