Low Form Borosilicate Glass Beakers
Actual product may vary
Low Form Borosilicate Glass Beakers
Low Form Borosilicate Glass Beakers
Low Form Borosilicate Glass Beakers
Low Form Borosilicate Glass Beakers
Low Form Borosilicate Glass Beakers
These low form beakers are made by hand from high quality borosilicate 3.3 glass. They are extremely durable, have low temperature gradients, and superior chemical durability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISS15441500 mL
FISS408895000 mL
Catalog number FISS15441
Price (SGD)
-
Capacity (Metric):
500 mL
  • Made from high quality borosilicate 3.3 glass
  • White graduations for easy visibility (not available for 5 mL and 10 mL sizes)
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Superior chemical durability
  • Low form
  • Autoclavable at 121°C
  • In compliance with DIN 12231 and ISO 3819 standards
Specifications
Capacity (English)16.9 oz.
ColorClear
DescriptionChemical durability
Diameter (English) Outer3.26 in.
Diameter (Metric) Inner80.5 mm
Diameter (Metric) Outer83 mm
Capacity (Metric)500 mL
MaterialBorosilicate Glass
For Use With (Application)Solution Preparation
Height (English)4.5 in.
Height (Metric)11.43 cm
Length (Metric)8.25 cm
Quantity1
TypeGlass Beakers
Width (English)3.25 in.
Width (Metric)8.25 cm
SterilityNon-sterile
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
ScaleWhite Graduated Scale
StyleLow Form
Unit SizeEach