Polyethylene Wash Bottles, LDPE
Polyethylene Wash Bottles, LDPE
Actual product may vary
Polyethylene Wash Bottles, LDPE
Cap fit with flexible polyethylene delivery tube which can be aimed at wherever required.
Have Questions?
Catalog NumberCapacity (Metric)
FISS14093500 mL
Catalog number FISS14093
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
  • Made from polyethylene
  • Translucent and unbreakable
Specifications
Capacity (Metric)500 mL
Capacity (English)17 oz.
ColorClear
MaterialPolyethylene
SterilityNon-sterile
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeWash Bottle
Chemical LabelUnlabeled
Unit SizeEA