Premium Museum Grade Dissection Pins
Premium Museum Grade Dissection Pins
Actual product may vary
Premium Museum Grade Dissection Pins
Make dissection easy with the proper tools. The Eisco™ Premium Museum Grade Dissection Pins are collection/museum grade insect entomology dissection pins.
Have Questions?
Catalog NumberSize
FISS13976Size 1
Catalog number FISS13976
Price (SGD)
-
Size:
Size 1
  • Premium, museum grade dissection pins
  • Double black enamel coated with nylon gold heads
  • Handmade of spring steel
  • Length: 38mm
Specifications
Product TypeDissection Needles
Blade MaterialSpring Steel
AutoclavableNot Autoclavable
CoatingBlack enamel
ColorBlack
MaterialNylon Gold Head
Quantity100 Pk.
SizeSize 1
Length (English)1.7 in.
Length (Metric)4.31 cm
Length (Metric) Blade38 mm
Diameter (Metric) Blade0.40 mm
Diameter (Metric) Face38 mm
Diameter (Metric) Head0.40 mm
Weight (Metric)45.35 g
Unit SizePK