Porcelain Crucible with Lid, Tall Form
Porcelain Crucible with Lid, Tall Form
Actual product may vary
Porcelain Crucible with Lid, Tall Form
Used to heat chemical compounds. EISCO Porcelain Crucibles are standard lab crucibles and can withstand temperatures up to 1050°C.
Have Questions?
Catalog NumberCapacity (Metric)Quantity
FISS1395450 mL10/Pk
Catalog number FISS13954
Price (SGD)
-
Capacity (Metric):
50 mL
Quantity:
10/Pk
  • Crucible for basic laboratory use to heat chemical compounds
  • Tall form body with lid
  • Constructed of glazed porcelain for strength, thermal performance, and enhanced chemical resistance
  • Can withstand temperatures up to 1050°C (1900°F)
Specifications
MaterialPorcelain
Max. Temperature (Metric)1050°C
Quantity10/Pk
Capacity (Metric)50 mL
AutoclavableAutoclavable
Diameter (Metric) Outer51 mm
Height (Metric)44 mm
Product TypeCrucible with lid
PorosityMedium
ShapeConical
Unit SizePK