Premium Three Finger Clamp w/ Dual Screw Adjustments
Actual product may vary
Premium Three Finger Clamp w/ Dual Screw Adjustments
Premium Three Finger Clamp w/ Dual Screw Adjustments
Premium Three Finger Clamp w/ Dual Screw Adjustments
Premium Three Finger Clamp w/ Dual Screw Adjustments
Premium Three Finger Clamp w/ Dual Screw Adjustments
Premium Three Finger Clamp w/ Dual Screw Adjustments
Choose durable lab equipment that will last for years. The EISCO Premium Three Finger Clamp w/ Dual Screw Adjustments is an industrial three finger, rubber-coated extension lab clamp with dual screw adjustments.
Have Questions?
Catalog NumberColor
FISS13911Blue
Catalog number FISS13911
Price (SGD)
-
Color:
Blue
  • Heavy duty spring loaded clamp jaw with dual screw adjustment to fine tune any fitting
  • Made of steel and powder coated alloy with all metal adjustment screws
  • 3 rubber coated clamp fingers
  • 6cm maximum opening clamp
  • 12.5cm steel connecting rod
Specifications
TypeClamp
MaterialSteel, Powder-Coated Alloy, Metal (Screws)
Size (English) Grip6 cm max. opening
ColorBlue
Jaw Type3-Prong
Unit SizeEach