Porcelain Evaporating Dishes
Porcelain Evaporating Dishes
Actual product may vary
Fisherbrand
Porcelain Evaporating Dishes
Designed to withstand high temperatures with proper use. GSC International Porcelain Evaporating Dishes can withstand temperatures of up to 1050°C on their glazed portions, and up to 1350°C on their unglazed bottom surfaces.
Have Questions?
Catalog NumberCapacity (Metric)
FISS13781200 mL
Catalog number FISS13781
Price (SGD)
-
Capacity (Metric):
200 mL
  • Made from high quality, laboratory grade porcelain
  • Uniform wall thickness and dense materials provide stable mechanical strength and little permeability
  • When used properly, glazed pieces can withstand temperatures of 1050°C and unglazed pieces can withstand temperatures up to 1350°C
  • Glazed inside and outside except for the bottom surface
Specifications
Capacity (Metric)200 mL
MaterialPorcelain
ShapeRound
TypeEvaporating Dish
Interior FinishGlazed
Temperature Range (Metric)1050°C
Unit SizePK