Wire Gauze with Ceramic Center
Actual product may vary
GSC
Wire Gauze with Ceramic Center
Distributes heat evenly while using a Bunsen burner. GSC International Wire Gauze with Ceramic Center features rolled smooth wire edges and a clean white center ceramic. A variety of sizes are available.
Have Questions?
Catalog NumberDimensions (L x W)
FISS137325 x 5 in.
Catalog number FISS13732
Price (SGD)
-
Dimensions (L x W):
5 x 5 in.
  • Wire gauze is used with Bunsen burners to aid in the distribution of heat
  • The ceramic center helps prevent flame from coming into direct contact with the container being heated
  • Helps protect lab equipment
  • Features rolled smooth wire edges and clean white center ceramic
Specifications
DescriptionRolled smooth wire edges
Dimensions (L x W)5 x 5 in.
For Use With (Application)Distribution of Heat with Bunsen Burners
Length (English)5 in.
Width (English)5 in.
MaterialCeramic
TypeWire Gauze
Unit SizePack