Clay Triangle
Actual product may vary
GSC
Clay Triangle
The perfect tool for suspending a crucible. The GSC Clay Triangles are made for heating crucibles. These clay triangles are made with galvanized iron wires to help prevent premature rusting.
Have Questions?
Catalog NumberLength (English)Length (Metric)
FISS137302 in.5.1 cm
Catalog number FISS13730
Price (SGD)
-
Length (English):
2 in.
Length (Metric):
5.1 cm
  • Great for heating crucibles
  • Made with galvanized iron wires
  • Rust resistant
  • Note: The length shown in the specifications is the length of the sides
Specifications
Length (English)2 in.
Length (Metric)5.1 cm
Unit SizePack