Reversible Rack for Microcentrifuge Tube
Reversible Rack for Microcentrifuge Tube
Actual product may vary
Fisherbrand
Reversible Rack for Microcentrifuge Tube
Reversible Rack for Microcentrifuge Tube made of polypropylene.
Have Questions?
Catalog NumberPositionsArrayLength (Metric)Width (Metric)
FISS130969612 x 8220 mm150 mm
Catalog number FISS13096
Price (SGD)
-
Positions:
96
Array:
12 x 8
Length (Metric):
220 mm
Width (Metric):
150 mm
  • Autoclavable
  • Molded-in coding for easy identification of tubes
  • Contains grids on both sides
  • One side holds 1.5 to 2 mL micro centrifuge tubes with extra smaller positions for 0.5 mL tubes
  • Other side holds 0.5 mL micro centrifuge tubes with extra positions for 1.5 to 2 mL micro centrifuge tubes
  • 96 place rack contains 12 x 8 arrays
  • 80 place rack contains 16 x 5 arrays
Specifications
Product TypeMicrocentrifuge Rack
Array12 x 8
AutoclavableAutoclavable
ColorBlue
HoldsMicrotubes (0.5 and 1.5/2 mL)
MaterialPolypropylene
Positions96
ShapeRectangular
ShelvesNone
StackableNo
SterilityNon-sterile
For Use With (Equipment)Microcentrifuge Tube
Quantity6 Pk.
Dimensions (L x W x H)220 x 150 x 50 mm (8.6 x 5.9 x 1.9 in.)
Length (Metric)220 mm
Length (English)8.6 in.
Width (Metric)150 mm
Width (English)5.9 in.
Height (Metric)50 mm
Height (English)1.96 in.
Unit SizePK