Test Tube Brush with Nylon Bristles
Actual product may vary
United Scientific Supplies
Test Tube Brush with Nylon Bristles
This Test Tube Brush has a straight brush top. For effective cleaning of test tubes in laboratories, these test tube brushes with nylon bristles are mounted in twisted heavy gauge galvanized wire.
Have Questions?
Catalog NumberLength (English)Length (English) BrushDiameter (English) Bristle
FISS1307011 in.4 in.1.5 in.
Catalog number FISS13070
Price (SGD)
-
Length (English):
11 in.
Length (English) Brush:
4 in.
Diameter (English) Bristle:
1.5 in.
  • Good quality nylon bristles.
  • Mounted in twisted heavy gauge galvanized wire.
  • All brushes feature a radial top.
Specifications
Product TypeTest Tube Brush
Bristle TypeRadial, Tufted End
Color-codedNo
Handle MaterialGalvanized
Handle TypeTwisted Wire
Tip TypeRadial Tip
ScentUnscented
SterilityNon-sterile
ColorWhite
Diameter (English) Bristle1.5 in.
Diameter (Metric) Bristle3.8 cm
For Use With (Equipment)Test Tubes
Length (English)11 in.
Length (English) Brush4 in.
Length (Metric)27.9 cm
Length (Metric) Brush10.2 cm
Size27.9 x 3.8 cm (11 x 1.5 in.)
Unit SizePack