White Light Box
Actual product may vary
White Light Box
White Light Box
Edvotek
White Light Box
Safely enhance the visualization of DNA stained with FlashBlue™, proteins stained with Coomassie Blue and autoradiograms with the Edvotek White Light Box.
Have Questions?
Catalog NumberDisplay
FISS11552None
Catalog number FISS11552
Price (SGD)
-
Display:
None
Safely enhance the visualization of DNA stained with FlashBlue™, proteins stained with Coomassie Blue and autoradiograms with the Edvotek White Light Box.
  • Features a spacious 25 x 25cm (9.94 x 9.84 in.) viewing area
  • Illuminated by long life LEDs
  • Housed in a slim aluminum body
  • Does not include an amber gel viewing filter
  • Power Rating: 8 Watts
  • Power Supply: uses external 110V power brick
Specifications
DisplayNone
Hertz50/60 Hz
Voltage110/220 V
Size25 x 25 cm viewing area
For Use With (Application)Gel Imaging
Product TypeGel Imaging System
Unit SizeEach