Porcelain Mortars and Pestles
Actual product may vary
Supertek Scientific
Porcelain Mortars and Pestles
Motor with spout.
Have Questions?
Catalog NumberCapacityDiameter (Metric) Inner
FISS09570B9.29 oz., 275mL10cm
Catalog number FISS09570B
Price (SGD)
-
Capacity:
9.29 oz., 275mL
Diameter (Metric) Inner:
10cm
Motor with spout.
  • Glazed outside except for bottom
  • Unglazed inside
  • Pestle unglazed to grinding surface
Specifications
Capacity (English)9.29 oz.
Diameter (English) Inner3.93 in.
Diameter (Metric) Inner10cm
Product TypeMortar and Pestle
MaterialPorcelain
For Use With (Application)Used to grind, crush and mix substances into fine powders
IncludesSpout
Capacity9.29 oz., 275mL
FinishGlazed Outer, Unglazed Inner
Capacity (Metric)275 mL
Unit SizeEach