Advanced Dissecting Kit for Biology Classes
Advanced Dissecting Kit for Biology Classes
Actual product may vary
Advanced Dissecting Kit for Biology Classes
Advanced Dissecting Kit for Biology Classes
Fisherbrand
Advanced Dissecting Kit for Biology Classes
Advanced Dissecting Kit for high school and college biology classes. Finest quality tools to provide ultimate dissection experience.
Have Questions?
Catalog number FISS06829
Price (SGD)
-
Advance dissecting kit is designed with fine-quality dissecting scissors, forceps, and screw-lock scalpel to provide the ultimate dissection experience. This dissecting kit also contains forceps that feature fine points and a guide pin for fine dissection work. Fine point scissors are very sharp and provide smooth cutting every time. This dissection kit also contains 6 extra blades for many dissections. Ideal for high school and college biology classes.
Kit Contains:
  • A scalpel with Screw Lock Blade (P/No: 21)
  • Six No.22 blades (P/No: B22)
  • Forceps 4.5 in. with Guide Pin, Fine Points (P/No: 15)
  • Iris Scissors, fine points, 4.5 in. (P/No: 9)
  • Dissecting Scissors with One Point Sharp and Other Point Blunt 5.5 in. (P/No: 5SB)
  • Teasing Needle - Straight (P/No: 31)
  • Teasing Needle - Angular (P/No: 32)
  • Mall Probe - Chrome 6 in. (P/No: 36)
  • Ruler 6 in. - Plastic - with inches and centimeters markings (P/No: 30)
  • Glass Dropper (P/No: 39)
  • Double-fold Vinyl Case (P/No: 55)
Specifications
Case MaterialVinyl
For Use With (Application)Finest quality tools to provide ultimate dissection experience
IncludesScalpel, 6 x No.22 Blades, Forceps 4.5 in., Iris Scissors 4.5 in., Dissecting Scissors, 2 x Teasing Needles, Chrome Mall Probe 6 in., Ruler 6 in., Glass Dropper, Double-fold Vinyl Case
Quantity1 Ea.
Product TypeAdvance Dissecting Kit
Unit SizeEA